Micro Essential Lab Hydrion™ Lo Iodine Test Kit
Provides a simple, reliable, and economical means to measure quarternary concentration of ammonium, chlorine and iodine sanitizer solutions
Supplier:  Micro Essential Lab IL250
Catalog No. 15-420-2C
Explore available promotions
Includes:
  • Hydrion Lo Ion Quat Test Kit: one 15-foot roll of test paper, matching color chart, two sample tubes with caps, an extra roll of test paper and a laminated color chart
  • Hydrion Quat Test Paper: each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart
Description
Description
Federal, State and Local health regulations require users of Quaternary Ammonium Sanitizer Solutions to have appropriate test kits available to verify the strength of sanitizer solutions. Sanitizer solutions are essential in the food service industry to ensure that sanitizers are at the proper concentration specified by the individual manufacturer.
Lo Iodine Test (FS Cat. No. 15-420-2C)
  • Estimate titratable iodine
  • Color matches at 0-12.5-25-50ppm
  • 15 ft. (4.6m) roll
  • 100 tests
Specifications
Specifications
To estimate the amount of titratable iodine in sanitizing solutions made with commercial iodophors
One 15 ft. roll of iodine test paper, matching color chart and two sample tubes with caps, color chart
100 Tests
Lo Iodine Test Kit