Micro Essential Lab Hydrion™ Quat Test Paper Refill
Test paper measures from 0 to 400ppm
Supplier:  Micro Essential Lab QTR10
Catalog No. 15-420-1E
Explore available promotions
Description
Description
  • Each pack comes with two calibrated color charts
  • Readings at 0, 100, 200, 300 and 400ppm
Specifications
Specifications
2 color charts
5 Rolls
Paper Refill