missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Micro Essential Lab Hydrion™ Sanitizer Test Kits: Quaternary Test Paper Kits

Provides a simple, reliable and economical means to measure quaternary concentration of ammonium, chlorine and iodine sanitizer solutions

Supplier:  Micro Essential Lab QT10

 View more versions of this product

Catalog No. 15-420-1A


Explore available promotions

Includes:
  • Hydrion Lo Ion Quat Test Kit: one 15-foot roll of test paper, matching color chart, two sample tubes with caps, an extra roll of test paper and a laminated color chart
  • Hydrion Quat Test Paper: each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart

Description

Description

Provides a simple, reliable and economical means to measure quaternary concentration of ammonium, chlorine and iodine sanitizer solutions

Federal, State and Local health regulations require users of Quaternary Ammonium Sanitizer Solutions to have appropriate test kits available to verify the strength of sanitizer solutions. Sanitizer solutions are essential in the food service industry to ensure that sanitizers are at the proper concentration specified by the individual manufacturer.

Hydrion Test for Two-chain Quats (FS Cat. No. 15-420-1A)

  • Measures 0 to 400ppm
  • Detects exhaustion of solutions that should be replaced, as well as helping to avoid using excessive amounts of sanitizing agents
  • Calibrated color chart
  • Color matches at 0-100-200-300-400ppm
  • Measures n-alkyldimethylbenzyl and/or n-alkyl dimethyl ethyl benzyl ammonium chloride and Roccall II
  • Each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart

Specifications

Specifications

To measure the concentration of quaternary sanitizers, including n-Alkyldimethylbenzyl and/or n-Alkyl dimethyl ethyl benzyl ammonium chloride and Rocall II
10 Pack
QT-10 Test for Two-chain Quats
One 15 ft. roll of test paper, matching color chart
0 to 400 ppm
Videos
SDS
Documents

Documents

Promotions

Promotions