MilliporeSigma™ Supelco™ Micro-Metering Valve (Low Pressure)
Designed for the control of fluid flow rates
$146.72 - $170.49
Specifications
For Use With (Application) Peristaltic Pumps, Pump Fluid-Transfer, Mass Spectrometry and Fraction Collection
Material PEEK
Type Low Pressure
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
111004880
View Documents
MilliporeSigma™ Supelco™
502367
1/4-28 Flangeless Fittings
Each for $146.72
111004777
View Documents
MilliporeSigma™ Supelco™
502375
10-32 Fingertight Fittings
Each for $170.49
Description
Description
  • Maximum Pressure: 800 psi
Specifications
Specifications
Peristaltic Pumps, Pump Fluid-Transfer, Mass Spectrometry and Fraction Collection
PEEK
Low Pressure
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'