Fisherbrand™ Micro Spatulas Set of 8
The versatile design of these stainless steel models enables use as a spatula, spoon, Scraper, or stirring rod. These multi-purpose items are rustproof and feature nonstick surfaces.
Catalog No. 13-820-086
Explore available promotions
Description
Description
Fisherbrand™ Assorted Micro Lab Spatula Set of 8. Most popular multipurpose spatulas for various applications
  • Ideal for transferring, collecting, mixing, or handling chemical or biological samples
  • Rustproof
  • Non-sticking
  • Stainless steel construction
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Specifications
8
Set of 8 Microspatula
Multiple uses
Stainless Steel
Autoclavable
Micro Spatula
5.5in.-8in.
13.97 to 20.32 cm