MilliporeSigma™ Supelco™ Micro-Splitter Valves
Available in High and Low Pressure options
Supplier: MilliporeSigma™ Supelco™ 502383
Specifications
Graduated | |
800 psi (Max) | |
3 Ports |
PEEK/PTFE | |
Low Pressure | |
1/4-28 Flangeless Fittings |