Integra™ Miltex™ Micro Surgery Scissors
Sharp points
Supplier: Integra™ Miltex™ 172110
Description
- High-quality surgical-grade stainless steel construction provides optimal performance
- Fully tempered precision screw locks maintain perfect alignment
- Ergonomic design combines weight, balance and feel for optimal comfort and performance
- Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
- Available in straight, curved and angled patterns
Specifications
Curved | |
7.125 in. | |
Stainless Steel | |
Sharp/Sharp |
Surgical | |
18 cm | |
Surgical, micro surgery |