Integra™ Miltex™ Micro Surgery Scissors
Sharp points
Supplier:  Integra™ Miltex™ 172150
 View more versions of this product
Catalog No. 12-460-234
Explore available promotions
Description
Description
  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Available in straight, curved and angled patterns
Specifications
Specifications
Straight
15 cm
Sharp/Sharp
6 in.
Surgical, Micro Surgery