missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ Microcentrifuge Tube Racks

Designed to hold 1.5mL microcentrifuge tubes.Microcentrifuge Tube Racks are constrcuted of autoclavable polypropylene with an operating temperature range between -20°C to +90°C.

$111.25 - $437.22

Specifications

Product Type Microcentrifuge Tube Rack
Material Polypropylene
View More Specs

Products 3
Catalog Number Mfr. No. Color Positions Quantity Price Quantity  
Catalog Number Mfr. No. Color Positions Quantity Price Quantity  
13-883-15
View Documents
BrandTech™ BRAND™
4341052
Red 128 5/Pk. N/A
 
13-883-16
View Documents
BrandTech™ BRAND™
4341002
Red 84 5/Pk. N/A
 
13-883-06
View Documents
BrandTech™ BRAND™
780605
White 20 1 N/A
 
Description

Description

Designed to hold 1.5mL microcentrifuge tubes. BrandTech™ BRAND™ Microcentrifuge Tube Racks are constrcuted of autoclavable polypropylene with an operating temperature range between -20°C to +90°C. The 84-tube and 128-tube versions fit 1.5mL tubes and are stackable with alphanumeric markings, while the 20-tube version has numbered positions and will fit either 1.5mL tubes or 0.5mL tubes with an adapter.

  • Manufactured of autoclavable polypropylene (121°C, 2 bar; acc. DIN EN 285).
  • BRAND™ Microcentrifuge Racks are available in 20-, 84-, and 128-tube versions.
  • The 84-tube and 128-tube versions fit 1.5mL tubes, are stackable with alphanumeric markings
  • Operating temperature range -20°C to +90°C.
  • The 20-tube version has numbered positions, and will fit either 1.5mL tubes or 0.5mL tubes with an adapter (Cat. No. 780608).
  • Also holds microcentrifuge tubes from other manufacturers.
Specifications

Specifications

Microcentrifuge Tube Rack
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'