Ansell MICROCHEM™ by AlphaTec™ Overboots
Ansell™ MICROCHEM™ by AlphaTec™ Overboots are air and water vapor permeable overboots with Sure Step sole and elastic foot opening
Supplier: Ansell WH15S9242205
Description
Air and water vapour permeable (breathable) to help reduce the risk of heat stressSpecifications
12 to 14 | |
White | |
Elastic Opening | |
Inspection, Asbestos Related Work, Powder Handling | |
150/Cs. | |
Boot Covers |
SMS | |
Single-use | |
Serged | |
EN 1073-2:2002; Type 5 EN ISO 13982-1:2004 + A1:2010; Type 6 EN 13034:2005 + A1:2009 | |
48 gsm |