missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff
Disposable nitrile gloves with extended cuff provide durable protection in the event of chemical splashes. NFPA and medical exam grade certified for use by healthcare professionals.
$33.33 - $35.36
Specifications
Certifications/Compliance | NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600 |
---|---|
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, EN 455 1-4 |
Applications | Healthcare, Laboratory |
Color | Black |
Disposable | Single-use |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-500-804
|
Ansell
93862070 |
Small |
Pack of 100 Each for $33.33
Case of 10 Pack for $333.30
|
|
|||||
19-500-805
|
Ansell
93862080 |
Medium |
Pack of 100 Each for $33.33
Case of 10 Pack for $333.30
|
|
|||||
19-500-806
|
Ansell
93862090 |
Large |
Pack of 100 Each for $33.33
Case of 10 Pack for $333.30
|
|
|||||
19-500-807
|
Ansell
93862100 |
X-Large |
Pack of 100 Each for $33.33
Case of 10 Pack for $333.30
|
|
|||||
19-500-808
|
Ansell
93862110 |
2X-Large |
Pack of 100 Each for $35.36
Case of 10 Pack for $353.60
|
|
|||||
Description
- Tested against both fentanyl and gastric acid (vomit) to simulate authentic overdose situations
- Dark, black color makes it easy to see when hands have come into contact with potentially hazardous powders
- Made with a soft, flexible nitrile formulation
- Fully textured for a reliable, consistent grip
- For use with:
- Administering drugs
- Emergency services
- Equipment repair and maintenance
- Extra protection over wrist and forearm
- Inspection, selecting, checking parts
- Laboratory analysis and testing
- Paint and spray shops
- Patient examinations
- Raw material sample collection
- Tire rotation and change
Specifications
NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600 | |
Healthcare, Laboratory | |
Single-use | |
6.3 mil | |
Yes | |
4.7 mil | |
Powder Free | |
0.16 mm | |
12 in. |
EN 420:2003 + A1:2009, EN 455 1-4 | |
Black | |
Fully Textured | |
0.16 mm | |
Nitrile | |
0.12 mm | |
6.3 mil | |
MICROFLEX™ MidKnight™ XTRA 93-862 | |
30 cm |