Carson™ MicroFlip Pocket Microscope with Flip Down Slide Base and Smartphone Digiscoping Clip
Carson™ MicroFlip Pocket Microscope features a flip-down slide base and smartphone digiscoping adapter clip.
Supplier: Carson™ EPDMP250
Description
- Contains a unique flip-down slide base
- Magnification ranges from 100X to 250X power.
- Built-in UV flashlight that is ideal for detecting counterfeit currency, nectar mapping and more.
- Comes with a smartphone digiscoping clip, 1 starter slide with cover slip and a wrist strap for convenient portability.
- Use the smartphone digiscoping clip to take pictures and record video using the built-in camera application on a smartphone
- Digiscoping clip will work with most smartphone models in the marketplace including iPhone, Samsung, HTC, LG and more.
- Thicker cases like Otterbox style cases may need to be removed before attaching the digiscoping clip.
- Powered by 1 AA battery (not included)
- Excellent educational field microscope for multiple hobbies, tasks, learning and exploration.
- For kids, students and even professional usage.
- Lifetime limited warranty
- Fits all phones
Specifications
Flip Down Slide Base and Smartphone Digiscoping Clip | |
Microscope | |
LED and UV | |
100 to 250x |