Learn More
Thermo Scientific™ Microlite™ White Microtiter™ Plates
Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Microlite Microtiter Plates.
$842.61 - $842.61
Specifications
Binding Property | Slightly Hydrophilic |
---|---|
Detection Method | Chemiluminescence |
Color | White |
Product Type | Microplate |
Material | Polystyrene |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
14-245-181
|
Thermo Scientific™
741712 |
Plate; Microlite 2 |
Case of 50 Each for $842.61
|
|
|||||
Description
Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Thermo Scientific™ Microlite White Microtiter Plates.
Available in plates or 12-well breakable strips.
- Polystyrene
- Choice of two surfaces, each performance assayed using an IgG binding assay
SURFACES
Microlite 1+ - Hydrophobic surface for binding of biomolecules with hydrophilic domains
Microlite 2+ - Slightly hydrophilic surface for increased binding of biomolecules with hydrophilic/hydrophobic character such as many proteins
Note:
Other configurations are available in Microlite 1 and 2 formulations.
Specifications
Slightly Hydrophilic | |
White | |
Polystyrene | |
None |
Chemiluminescence | |
Microplate | |
Non-sterile | |
Antibodies (Glycoproteins) and Proteins |