missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences MicroLiter 11 mm Crimp Top Autosampler Vials

MicroLiter 11 mm Crimp Top Autosampler Vials are cleaned and packaged in a certified Class 10,000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials. Vials and inserts meet strict tolerances.

Supplier:  DWK Life Sciences W111400

 View more versions of this product

Catalog No. 15-111-565


Explore available promotions

Description

Description

For use with Microliter 11 mm Crimp Top Autosampler Vial seals
Specifications

Specifications

Clear
Borosilicate Glass (Type 1)
100
32 mm
No
12 mm
1.8 mL
MicroLiter
11 mm
Videos
SDS
Documents

Documents

Promotions

Promotions