missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences MicroLiter 9 mm Screw Thread Polypropylene Autosampler Vials

MicroLiter 9 mm autosampler polypropylene screw thread vials are identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results.

Supplier:  DWK Life Sciences 0913001000

 View more versions of this product

Catalog No. 15-111-328


Explore available promotions

Description

Description

Performance equal to an 11 mm crimp-top vial. Inserts available.
Specifications

Specifications

Clear
Polypropylene
100
32 mm
No
12 mm
1 mL
MicroLiter
9 mm
9 mm Screw Thread Caps - MicroLiter
Videos
SDS
Documents

Documents

Promotions

Promotions