missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.
Supplier: Greiner Bio-One 705073
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Individual wells can be pipetted without leaving residue
- Flat bottom enables precise optical measurements
- Suitable for transmission measurements
- Specifica
Specifications
Single break | |
Strip Plate | |
High Binding | |
Polystyrene | |
Non-sterile | |
C | |
Colorimetric immunoassay | |
Yes |
High Binding | |
8 x 12 | |
Clear, Red color coding | |
96 | |
Microlon | |
Yes | |
100/Cs. | |
127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.) |