Walter Stern Micrometer Calipers
$135.88 - $135.88
Specifications
Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
12-125
View Documents
Walter Stern
601A
Each for $135.88
12-126
View Documents
Walter Stern
601B
Each for $135.88
Description
Description
Metric Scale. 0 to 25mm range with metric micrometer calipers. Friction stop causes adjustment knob to slip at the same pressure each time, ensuring reproducibility. Reads to 0.01mm.
English Scale. Useful over 0 to 1 in. (0 to 25mm) range, readable to 0.001 in. (0.025mm). Otherwise similar to metric-scale version.
Specifications
Specifications
0.0
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'