Eppendorf™ Assay/Reader Microplate
Eppendorf Assay/Reader Microplates are designed for diverse microplate assays, optimizing absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
Supplier:  Eppendorf™ 951040196
 View more versions of this product
Catalog No. 05414244
Explore available promotions
Description
Description
  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples.
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximizing reflection.
  • Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes.
  • All plates are optimized for minimal autofluorescence and autoluminescence.
  • Available in lot-certified PCR clean purity grade.
  • Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability.
Specifications
Specifications
Autoclavable
96
6000 x g
White
Flat Bottom (F-well)
PCR Clean (Non-sterile)