FEATHER Microtome Blades
FEATHER Microtome Blades are specially coated for superior cutting performance. FEATHER's expanded microtome blade product line enables you to select the blade that best meets your needs.
$96.08 - $220.66
Specifications
Product Type Microtome Blade
Disposable Single-use
View More Specs
Products 9
Catalog Number Mfr. No. Description For Use With (Equipment) Cutting Angle Price Quantity  
Catalog Number Mfr. No. Description For Use With (Equipment) Cutting Angle Price Quantity  
36-100-5624
View Documents
FEATHER
A22
Excellent for soft tissue and thin sectioning; Good for ribbon sectioning Sledge and Rotary Microtome 22°
Pack of 50 Each for $220.66
36-099-2334 Edge
View Documents
FEATHER
A35
Excellent for hard tissue; Good for soft tissue, cryostat, and ribbon sectioning Sledge and Rotary Microtome 35°
Pack of 50 Each for $209.92
Edge
12-634-1F Edge
View Documents
FEATHER
C35
Excellent for cryostat Cryostat 35°
Pack of 20 Each for $96.08
Edge
36-099-2335
View Documents
FEATHER
HIGHPROFILE
Excellent for soft tissue and ribbon sectioning; Good for hard tissue Sledge and Rotary Microtome 35°
Pack of 50 Each for $214.95
36-099-667
View Documents
FEATHER
N35
Excellent for hard tissue; Good for soft tissue, thin sectioning, and ribbon sectioning Sledge and Rotary Microtome 35°
Pack of 50 Each for $213.71
12-631R Edge
View Documents
FEATHER
R35
Excellent for hard tissue and ribbon sectioning; Good for soft tissue Sledge and Rotary Microtome 35°
Pack of 50 Each for $201.95
Edge
12-631P
View Documents
FEATHER
S22
Excellent for soft tissue and extremely thin sectioning Sledge and Rotary Microtome 22°
Pack of 50 Each for $207.63
12-634-1C Edge
View Documents
FEATHER
S35
Excellent for soft tissue; good for cryostat and thin sectioning Sledge and Rotary Microtome 35°
Pack of 50 Each for $196.42
Edge
12-631C
View Documents
FEATHER
S35L
Excellent for soft tissue and large blocks Sledge Microtome 35°
Pack of 20 Each for $201.68
Description
Description
FEATHER's expanded microtome blade product line enables you to select the blade that best meets your needs:
  • A-22
    • For extra thin sectioning of paraffin embedded tissue blocks
    • Especially suited for fibered tissues and biopsy
  • A-35
    • For thin sectioning of paraffin embedded tissue blocks
    • Especially suited for hard tissue block and ribbon sectioning
  • C-35
    • For thin cryo-sectioning
    • Especially ground for cryostat use to eliminate curling or wrinkling problems
  • High-Profile
    • For thin sectioning of paraffin embedded tissue blocks
    • Especially suited for ribbon sectioning with rotary microtome
    • Standard blade for routine, easy to get ribbon sections
  • N-35
    • For thin sectioning of paraffin embedded tissue blocks
    • Especially suited for hard tissue; features long blade life with no need to move the blade in real sectioning after trimming
  • R-35
    • For thin sectioning of paraffin embedded tissue blocks
    • Especially suited for routine microtomy operations
    • Special hardened blade edge; double spattering increases edge strength and durability even when sectioning right after trimming with the same blade position
  • S-22
    • For extra thin sectioning of paraffin embedded tissue blocks
    • Especially recommended for laboratory and research use
    • Blade life may be limited under some circumstances because of the extra thin edge
  • S-35
    • For thin sectioning of paraffin embedded tissue blocks
    • Especially suited for routine microtomy operations and biopsy
    • Standard blade for routine sectioning with good sectioning quality including initial sectioning
  • S-35L
    • For thin sectioning of large paraffin embedded tissue blocks
    • Same quality as the S-35 with greater length (120 mm)
Specifications
Specifications
Microtome Blade
Single-use
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'