Learn More
Poltex Microtube Box Freezer Rack
A freezer rack that is suitable for -20°C freezer storage of small microtube storage boxes.
Supplier: Poltex FZRCK54
Description
- Durable 1/4 in. thick acrylic construction.
- Easily customizable to your specific setup.
Specifications
| Freezer Rack | |
| Acrylic | |
| 20 x 4.6 x 9.2 in. |
| Clear | |
| Yes |