Brady™ Mild Sheet Steel Portable Metal Group Lock Box
Simplifies lockout of large equipment
Supplier:  Brady™ 65699
 View more versions of this product
Catalog No. 18-999-1335
Explore available promotions
Description
Description
  • Heavy steel lock box stores keys after machine or process is locked out
  • Each authorized work team member adds a personal lock or tagout device on the group lock box
  • Ensures that no one employee has access unless all employees have removed their locks or tags
  • Accepts up to 13 individual locks
Specifications
Specifications
Red
1 Ea.