Honeywell™ Miller™ QuickPick™ Rescue Kit
Rescue Kits deliver peace of mind during unexpected, stressful peer-rescue situations
Supplier: Honeywell™ QP100FT
Description
The rescuer can remotely attach the system to the suspended worker while remaining securely anchored on the walking/working surface. All rescue components are contained in one easy-to-store kit with no assembly necessary — only requires an overhead anchorage.
- Out-of-the-bag solution for prompt rescue, with no assembly required
- Quickly raise or lower a fallen worker to safety — avoid suspension trauma
- Professional rescue solution; enhances worker safety and corporate rescue plan
Applications:
Construction, Manufacturing, Municipal Services, Oil and Gas, Transportation, Utilities, Wind Energy
Compliance
Meets ANSI Z359.4 requirements.
Specifications
20.5kg | |
QuickPick Premium Kit | |
181.4 kg | |
400 lb. |
100 ft. | |
45.4 lb. | |
OSHA, ANSI A10.32, ANSI Z359.4 | |
30.48 m |