Honeywell™ Miller™ Duralite Fall Arrest and Retrieval Harness and Positioning Harness
Full-body harnesses in lightweight, durable, yellow or gold polyester webbing. Cat. No. 18-542 is suitable for fall arrest and retrieval. Cat. No. 18-543 has all the features of 18-542, plus side D-rings for positioning applications.
Supplier:  Honeywell™ 8507UYKC
 View more versions of this product
Catalog No. 18-543
Explore available promotions
Description
Description
  • Lightweight but durable harnesses provide maximum worker mobility when job requires constant climbing
  • Mating leg buckles and color-coded leg straps make harness easy to put on and give a custom-fit feel
  • High-visibility 1-3/4 in. (4.4cm) yellow nylon webbing
  • Forged sliding full-circle back D-ring
  • Friction shoulder buckle
Specifications
Specifications
Fall Arrest and Retrieval
Sliding Back, Side