missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Miller MightEvac Self Retracting Lifeline Unit with Emergency Retrieval

Honeywell Miller MightEvac Self Retracting Lifeline Unit combines the performance and reliability of the MightyLite Self-Retracting Lifeline Series with a quick-activating retrieval mechanism for emergency evacuation.

$6037.39 - $6037.39

Specifications

Capacity (English) 310 lb.
Capacity (Metric) 140.62 kg
Color Red
Product Type Self-retracting Lifeline
For Use With (Application) Fall Protection
View More Specs

Products 1
Catalog Number Mfr. No. Length (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Price Quantity  
19-085-479
View Documents
Honeywell
MR50GBZ750FT
50 ft.
Each for $6,037.39
 
Description

Description

  • No annual factory recertification required– Significant long-term savings/lower cost of ownership
  • 20% lighter than competitive models
  • Promotes quick and easy rescue with a 4:1 mechanical advantage (average speed of 25 ft. [7.6m] per minute)
  • Corrosion-resistant stainless steel/aluminum components
  • Convenient, heavy-duty carrying handle
  • 310 lb. (1.4kN) weight capacity
Specifications

Specifications

310 lb.
Red
Fall Protection
140.62 kg
Self-retracting Lifeline
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'