Honeywell™ Miller™ Revolution™ Harnesses
Design based on key user-identified needs
Supplier:  Honeywell™ RDTTBUBKU
 View more versions of this product
Catalog No. 19-130-4081
Explore available promotions
Description
Description
  • PivotLink™ connection with unique rotary design promotes greater comfort in bending/mobility; also serves as an attachment point for belts, tools and accessories as part of an integrated accessory system
  • Cam buckle provides easy, one-handed adjustment/release and allows simultaneous adjustment of shoulder straps
  • Semi-flexible ErgoArmor™ back shield minimizes impact from sharp/heavy snap hooks and self-retracting lifelines; knitted mesh membrane minimizes moisture absorption and maintains optimal air circulation
  • Stand-up back D-ring simplifies connection
  • DualTech™ webbing makes donning easy with two-sided, contrasting-color webbing and textures
  • High-impact-resistant polymer web finials provide positive gripping surface for easy web adjustment
  • Integrated self-contained label pack encapsulates labels, decreasing damage and loss
Order Info
Webbing tested to 7000 lb. tensile strength. Universal size.
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; CSA Z259.10 requirements.
Specifications
Specifications
Revolution Harness
Construction, Fall Protection
Standard Harness
Tongue-Buckle Leg Straps
Universal