MilliporeSigma™ Milli-Q™ IQ 7000 Ultrapure Water System
Work confidently with Milli-Q™ IQ 7000. The Milli-Q™ IQ 7000 system combines advanced technologies, hydraulic design and software capabilities to efficiently produce the superior-quality ultrapure water you need to work confidently.
Supplier:  MilliporeSigma™ ZIQ7000T0C
Catalog No. ZIQ7000T0
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Work confidently with Milli-Q™ IQ 7000. The Milli-Q™ IQ 7000 system combines advanced technologies, hydraulic design and software capabilities to efficiently produce the superior-quality ultrapure water you need to work confidently.
IQ 7000 Purification System. An ultrapure water solution designed with YOU in mind.
Simple and intuitive dispensing
  • Just a twist of the new dispensing wheel lets you deliver water at the flow rate you need, from precise drop by drop up to a full 2 liters per minute. For added efficiency, you can also directly select a dispensing option from the Home screen
  • While dispensing, you can easily check the water quality parameters, always visible onscreen. Their values are continuously monitored by the system, so you can have full confidence in your water quality.
Complementary technologies
  • The Milli-Q™ IQ 7000 system combines advanced technologies, hydraulic design and software
    capabilities to efficiently produce the superior-quality ultrapure water you need to work confidently.
A powerful combination of purification media
  • The slim IPAK Meta™ and IPAK Quanta™ cartridges are designed to function as a pair. The cartridges use a combination of Jetpore™ mixed-bed ion-exchange resin and innovative IQnano™ ion-exchange media to provide exceptional performance
Water quality to match all your application needs
  • You’ll find that our range of POD-Paks is the perfect solution for your various applications.
    Each of the Application-Pak polishers targets specific types of contaminants and removes
    them right at the Q-POD™ dispenser
Versatile configurations for the best fit
Each lab is unique. The Milli-Q™ IQ 7000 system lets you optimize valuable lab space to help maximize your efficiency
Easy data management for a paperless environment
  • Powerful, integrated data management makes it so much simpler and faster to manage your water system data — no more log book or paper to archive! All data generated by the Milli-Q™ IQ 7000 is stored in the system memory
DIGITAL SERVICES SIMPLIFY SYSTEM CARE AND ASSURE PRODUCTIVITY
MyMilli-Q™ Remote Care is a unique online monitoring and service feature that provides:
  • Remote access to your system and its data
  • Automatic notifications, by email or SMS, in case of an alert or alarm
  • Faster service with remote diagnostics and repair possible
  • Simplified data management and advanced traceability that eases audit and accreditation processes
Order Info
Q-PODS are not included with this item. See ZMQSP0D02 for Q-POD 2 and ZIQP0D000 for Q-POD 1.
Specifications
Specifications
0.055μS/cm
No particles with size >0.22 μm (with Millipak filter)
Pretreated water using EDI, DI, RO or distillation technologies
Up to 2 L/min.
Yes
≤2 ppb (in the appropriate operating conditions, otherwise typically <5 ppb)
Water Purification System
US
5°C to 35°C
35.3 lb.
100/240 V
Milli-Q IQ 7000 Ultrapure Water System. The most advanced Milli-Q ultrapure water system that is intelligent, intuitive and green.
1 to 250 L
Pure
Type 1
18.2 Mohm.cm
<5 ppb
<5 pg/mL (with Biopak polisher)
<1 pg/mL
13.8 x 10.4 x 19.6 in.
16.0 kg
50/60 Hz