missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Millicell™ Hanging Cell Culture Insert
Polyethylene Terephthalate hanging cell culture insert used in a 24-well plate.
Supplier: MilliporeSigma™ PTHT24H48
Description
- Polyethylene Terephthalate hanging cell culture insert with a pore size of 0.4 μm.
- Used in a 24-well plate for cell attachment, cell culture, cell differentiation and ICC.
- Replacement for MCHT24H48
Specifications
Replacement for MCHT24H48, Polyethylene Terephthalate hanging cell culture insert with pore size of 0.4 μm used in a 24-well plate for cell attachment, cell culture, cell differentiation & ICC. | |
PET | |
200 μL | |
8.0 μm | |
24-Well Culture Plates | |
48 (Individually Blister Packaged) | |
Sterile |
0.3 cm2 | |
400 μL | |
2 x 105 pores/cm2 | |
Cell Culture Insert | |
6.5 mm | |
Yes | |
24 |