Integra™ Miltex™ Olsen-Hegar Needle Holder
With suture scissors
Supplier:  Integra™ 816TC
 View more versions of this product
Catalog No. 12-460-569
Explore available promotions
Description
Description
Specifications
Specifications
Olsen-Hegar Needle Holder
Suture scissor
2600 Teeth/sq. in.
16.5 cm
Tungsten Carbide
Serrated jaws w/tungsten carbide cutting edges
6.5 in.