missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mineral Oil, Light (NF/FCC), Fisher Chemical™
Saybolt viscosity: 158 maximum
Supplier: Thermo Fisher Scientific O1211
Specifications
Mineral Oil, Light | |
Pass Test (NF) | |
White | |
260°C | |
Glass Bottle | |
MFCD00131611 | |
0.818 to 0.880 (FCC/NF) | |
Meets Requirements (NF) | |
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
452.363 | |
3 CS min. (FCC), 3.0 to 34.4 mm2.S-1 (NF) | |
FCC/NF |
8042-47-5 | |
Pass Test (FCC) | |
0.83g/mL | |
160°C | |
C16H10N2Na2O7S2 | |
1 L | |
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g | |
AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate | |
9566064 | |
<0.1 mmHg | |
Liquid |
Chemical Identifiers
8042-47-5 | |
452.363 | |
AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
9566064 | |
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
C16H10N2Na2O7S2 | |
MFCD00131611 | |
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g | |
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:0
Flammability:1
Instability:0