Cytiva Mini Dialysis Kits
Offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
$566.44 - $566.44
Specifications
Product Type | Mini Dialysis Kit |
---|---|
Kit Contents | 50 x tube with dialysis cap, 50 x standard cap, 6 x float |
Quantity | Each |
For Use With (Application) | Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane |
Includes: 50 tubes with 50 dialysis and 50 standard caps, plus six floats
Catalog Number | Mfr. No. | MWCO | Capacity (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | MWCO | Capacity (Metric) | Price | Quantity | ||||
45-001-736
|
Cytiva
80648432 |
8 kDa | 2 mL |
Each for $566.44
|
|
||||
45-001-733
|
Cytiva
80648375 |
1 kDa | 250 μL |
Each for $566.44
|
|
||||
45-001-734
|
Cytiva
80648394 |
1 kDa | 2 mL |
Each for $566.44
|
|
||||
45-001-735
|
Cytiva
80648413 |
8 kDa | 250 μL |
Each for $566.44
|
|
||||
Description
- Especially well-suited for desalting protein samples prior to analysis by isoelectric focusing (IEF) and 2-D electrophoresis
- Uses disposable dialysis tubes with dialysis membrane incorporated into caps
- Conical tube bottom maximizes sample recovery
- Process complete in as little as several hours
- Choice of molecular weight cut-off and tube size
Specifications
Mini Dialysis Kit | |
50 x tube with dialysis cap, 50 x standard cap, 6 x float | |
Each | |
Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane |