BUCHI Mini-DIN to RJ45 Communication Cable
For the connection between vacuum controllers and pumps and/or recirculating chillers.
Supplier:  BUCHI 11060649
Catalog No. 05405055
Explore available promotions
Description
Description
Provides Connections For:
  • Vacuum Controller V-850/V-855 to Vacuum Pump V-300/V-60
  • Vacuum Controller V-850/V-855, Interface I-100 to Recirculating Chiller F-305/F-308/F-3140
Specifications
Specifications
Communication Cable
Connection Between Vacuum Controllers and Vacuum Pumps
V-850, V-855, V-300,V-600, I-100, F-305, F-308, F-314