missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ Mini Tube System
Polypropylene tube system for compound library storage
Supplier: Axygen™ MTS06C
Description
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
- Rack features a one-way lid design and alpha-numeric grid
- Molded-in condensation rings on the lid minimize cross contamination
Specifications
Autoclavable | |
Clear | |
Ensures positive sample identification | |
960/Pk. | |
Non-sterile |
0.65 mL | |
Polypropylene | |
Alpha numeric grid, one way lid | |
960 Tubes/Bag, 5 Bags/Cs. | |
Round |