Axygen™ Mini Tube System
Polypropylene tube system for compound library storage
Supplier: Axygen™ MTS8CPC
Description
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
- Rack features a one-way lid design and alpha-numeric grid
- Molded-in condensation rings on the lid minimize cross contamination
Specifications
8 Cap Strip | |
Polyethylene | |
120/Pk. | |
Non-sterile |
Clear | |
Ensures positive sample identification | |
120 Strips/PK., 5 Packs/Cs. | |
Strip |