Supertek™ Miniature DC Motor DFS Item
Miniature DC mortor, a component part of many kits, this pre-assembled miniature motor operates on 1.5V or 3V DC.
Supplier:  Supertek™ PH502011S3
Catalog No. S012321
Explore available promotions
Description
Description
  • Miniature DC motor.
  • A component part of many kits.
  • This pre-assembled miniature motor operates on 1.5V or 3V DC.
  • RPM is between 7,000 to 11,000.
  • Comes with attached 12 in. red and black leads.
Specifications
Specifications
DC Motor