missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minigrip™ SPECI-ZIP™ Reclosable Clear Biohazard Bags with Black and Orange Biohazard Warning
Designed with temperature indicators and tear line
$105.35 - $217.10
Specifications
Product Type | Reclosable Biohazard Bag |
---|---|
Color | Clear |
For Use With (Application) | Easy-to-open tear line |
Includes | Black and orange biohazard warning, double pocket, temperature indicators and tear line |
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Height (English) | Height (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Height (English) | Height (Metric) | Price | Quantity | ||
22-310-402
|
Minigrip™
IP46B3T |
4 in. | 10.16 cm | 6 in. | 15.24 cm |
Case of 1000 Each for $105.35
|
|
||
22-310-403
|
N/A | 6 in. | 15.24 cm | 9 in. | 22.86 cm |
Case of 1000 Each for $217.10
|
|
||
22-310-405
|
N/A | 12 in. | 30.48 cm | 15 in. | 28.1 cm |
Case of 250 Each for $210.55
|
|
||
22-310-406
|
N/A | 20 in. | 50.8 cm | 24 in. | 60.96 cm |
Case of 100 Each for $190.45
|
|
||
Description
Designed with temperature indicators and tear line
- Double pocket, three-wall bags
- Bilingual in English and Spanish
Specifications
Reclosable Biohazard Bag | |
Clear | |
Easy-to-open tear line | |
Black and orange biohazard warning, double pocket, temperature indicators and tear line |