missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minuteman™ CRV Vacuum Accessories
Filters for use with CRV Cleanroom Vacuums
$149.11 - $149.11
Specifications
For Use With (Application) | Class 1 through Class 100,000 Cleanrooms Vacuuming |
---|
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Quantity | Price | Quantity | |||
18-999-283
|
Minuteman™
805044 |
Filter Cloth Bag Assembly | CRV and RFI/EMI vacuum Filtration Systems, Air Vacuum 6 gal. | Each |
Each for $149.11
|
|
|||
Specifications
Class 1 through Class 100,000 Cleanrooms Vacuuming |