missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Minuteman™ Lead Vacuum DFS Item

HEPA-filtered vacuum recovers lead, dust, paint chips, and other hazardous materials

Supplier:  Minuteman™ C8298506

Catalog No. 19-063-035


Explore available promotions

Includes:

50 ft. (15.2m) power cord, four 1.63 in. (4.14cm) casters, and a dry tool kit consisting of 1.25 in.W x 8 ft.L (3.18cm x 2.44m) crushproof hose, 1.25 in. (3.18cm) wide wand, 12 in. (30.5cm) wide crevice tool, 5 in. (12.7cm) wide upholstery tool, and 11 in. (27.9cm) combination floor/carpet tool.


Description

Description

Won't exhaust unseen lead particles back into the environment as many conventional vacuum cleaners do. Contains particles within a series of special filters, allowing no hazardous dust to escape back into the air.

  • Nonwoven, embossed polyester filter bag provides 50% more filter area than standard bag; 99.9% efficient at 1.0μm
  • HEPA filter is 99.97% efficient down to 0.3μm
  • Polypropylene lid assembly is acoustically designed to reduce noise of operation to 74dBA
  • Tank is constructed from durable, noncorrosive, dent-proof polyethylene
  • Sure grip/quick-release mechanism makes hoses quick and easy to connect and remove

Specifications

  • Voltage: 115VAC
  • Tank Size: 6 gal. (22L)
  • Power: 1.25hp
  • Airflow: 95cfm
  • Water Lift: 85 in. (21.6cm)
  • Dry Capacity: 0.46 cu. ft. (0.01m3)
  • Weight: 17 lb. (8kg)

Order Info

115VAC. Power: 1.25hp. Airflow: 95cfm. Water lift: 85 in. (21.6cm). Weight: 17 lb. (8kg).

Specifications

Specifications

115/220 V
50 ft. Power cord, Four 1.63 in. Casters, a Dry Tool Kit consisting of 8 ft. L x 1.25 in. W Crushproof Hose, 1.25 in. W Wand, 12 in. W Crevice Tool, 5 in. W Upholstery Tool, 11 in. Combination Floor/Carpet Tool
Lead Vacuum
0.01 m3/min.
768 oz.
HEPA
99%
85 in.
8 kg
17 lb.
0.34 cu. ft.
95 cfm
22 L
74 dB
1.25 hp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions