Minuteman™ Vacuum Accessories and Replacement Parts
Supplier: Minuteman™ 110029
Description
Impact filter can be used with both tank sizes. All other replacement parts are size specific. Plastic biohazard encapsulation bags assist in proper collection and disposal of potentially hazardous contaminants.
Specifications
Remediation of Hazardous Molds, Spores, Bio-haz Dust and Viral Particulate Containment | |
Each | |
ABS Plastic, Heavy Gauge Steel Mesh |
ULPA Replacement Filter | |
Bio-haz 6 gal. and 15 gal. and X-829 6 gal. and 15 gal., and Lead vacuum | |
99.99% |