Minuteman™ Vacuum Accessories and Replacement Parts DFS Item
Supplier:  Minuteman™ 800056
 View more versions of this product
Catalog No. 19-063-053
Explore available promotions
Description
Description
Impact filter can be used with both tank sizes. All other replacement parts are size specific. Plastic biohazard encapsulation bags assist in proper collection and disposal of potentially hazardous contaminants.
Specifications
Specifications
Remediation of Hazardous Molds, Spores, Bio-haz Dust and Viral Particulate Containment
Each
Cloth
Vacuum Filter
Bio-haz. 6 gal., X-829 6 gal., Lead Vacuum, X-839
Product Suggestions
Product Suggestions