missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minuteman™ X829 Series HEPA Vacuums
Safe, easy-to-use vacuum for difficult dry and wet cleaning jobs
Supplier: Minuteman™ C8291506
Description
Patented safety interlock system prevents operation if filter is incorrectly aligned.
- Removes hazardous dusts such as lead, arsenic, and asbestos, and nonhazardous wastes such as flour, wood, and foundry dusts
- Durable tank has a quick-release, self-sealing intake that ensures a positive vacuum seal
- ULPA (Ultra Low Penetration Air) filter is 99.999% efficient at 0.12μm; resists punctures, tears, leaks, abrasions, and moisture damage
- Patented safety interlock system ensures the ULPA filter is properly in place
- One-piece, polyethylene Uni-Adapter™ attaches easily to form a leakproof seal with the vacuum tank, enabling safe wet direct load recovery
Specifications
115/220 V | |
87 lb. | |
X829 series HEPA vacuum Wet or Dry | |
Wet: 13 gal and Dry: 0.82 cu. ft. | |
110 cfm | |
57 L | |
99.99% |
39 kg | |
Hazardous Dusts | |
Polyethylene | |
Wet: 49 L and Dry: 0.23 cu. m. | |
15 gal. | |
HEPA | |
105 in. |