missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ TransIT-PRO™ Transfection Kit
Specifically developed for mammalian protein production in suspension CHO and 293 cells. The PRO Boost Reagent is optional and enhances expression in certain media formulations. Use of TransIT-PRO Transfection Kit eliminates the need for a culture medium change post-transfection.
$1259.54 - $10087.29
Spécifications
| Content And Storage | -20°C |
|---|---|
| Cell Type | Mammalian |
| Format | Tube |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
MIR5700
|
Mirus Bio™
MIR5700 |
1 mL |
chaque for $1,259.54
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
MIR5760
|
Mirus Bio™
MIR5760 |
10 mL |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
- Suitable for both transient and stable transfection
- Compatible with multiple CHO media formulations
- Reproducible protein expression with minimal optimization
- Outperforms linear PEI in protein yield and provides a cost-effective alternative to FreeStyle™ MAX
Warranty
One year (when properly stored and handled).
Spécifications
| -20°C | |
| Tube | |
| Chemical Transfection | |
| Transfection Reagent |
| Mammalian | |
| None (Promoterless) | |
| Mammalian Protein production in suspension 293 and CHO derived cells, eliminates the need for a culture medium change post-transfection and is suitable for both transient and stable transfection | |
| Sterile |