Mirus Bio™ Ingenio™ Electroporation Cuvettes
Ingenio™ Electroporation Cuvettes for use with the Ingenio Electroporation System.
Supplier: Mirus Bio™ MIR50121
Description
- Deliver to hard to transfect cell lines and primary cells
- Use your existing system including Ingenio EZporator, Lonza-Amaxa, Bio-Rad or Harvard BTX
- Ingenio Electroporation Solution is available as a stand-alone solution or as part of a complete kit with cuvettes and cell droppers
Specifications
Sterile | |
Single-use | |
50 Cuvettes | |
0.2 cm | |
Electroporation |
Electroporation Cuvet | |
Polycarbonate | |
50 Pk. | |
Ingenio Electroporation Kit |