Learn More
Mitomycin C, Fisher BioReagents™
Supplier: Fisher BioReagents BP253120
Description
Mitomycin C
Inhibitor of DNA synthesis, nuclear division, and cancer cells. Antibacterial spectrum: Gram positive, gram negative, acid-fast bacilli.Specifications
Mitomycin C | |
360°C | |
≥98.0 % | |
C15H18N4O5 | |
15, 6300 | |
Soluble in water | |
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N | |
5746 | |
334.3 | |
Solid |
50-07-7 | |
Blue | |
Amber Glass | |
20 mg | |
mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c | |
NWIBSHFKIJFRCO-WUDYKRTCSA-N | |
334.332 | |
CHEBI:27504 | |
99.0% min. (RP CH3CN:H20 0 to 100%), 98.0% min. (RP MEOH:H20 90:10) |
Chemical Identifiers
50-07-7 | |
334.332 | |
mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c | |
CHEBI:27504 |
C15H18N4O5 | |
NWIBSHFKIJFRCO-WUDYKRTCSA-N | |
5746 | |
CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N |
Safety and Handling
WARNING!
Emergency Overview
Toxic if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is not required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:0
Instability:0
Recommended Storage : 4°C