Fisherbrand™ Mobile Lab Equipment Workstation, Phenolic Work Surface
Fisherbrand™ Mobile Lab Equipment Workstations are for labs needing a flexible and versatile equipment platform.
Description
- Can be positioned along the perimeter of a lab or placed back-to-back in an island configuration.
- They are furnished as complete assemblies that include the work surface and shelving.
- The steel shelving includes book end style brackets and a front and rear retaining lip.
- Shelves and work frames are adjustable in 1 in. increments.
- Workstations are furnished with vibration isolation leveling feet and 3 in. casters. Two are locking/swivel and two are swivel.
- Color is Fisherbrand White.
Specifications
2 tiers of shelving, Phenolic work surface, Vibration isolation leveling feet and casters | |
2 years | |
198.1 cm | |
408.2 kg | |
78 in. | |
White |
60 in. | |
30 in. | |
76.2 cm | |
152.4 cm | |
900 lb. |