MilliporeSigma™ Supelco™ Mobile Phase Reservoir System
This convenient system is made entirely of inert parts: glass, Tefzel, PEEK, and PTFE TFE. Includes 10 ft./3 m of 1/8 in. O.D. PTFE tubing, a PEEK bottom-of-the-bottle filter/sparger, a plug, and a 1-liter or 2-liter safety-coated bottle.
Supplier: MilliporeSigma™ Supelco™ SGMA55061
Specifications
HPLC | |
2 L |