Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
Supplier:  Epredia™ 88026
 View more versions of this product
Catalog No. 22-110-652
Explore available promotions
Description
Description
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
Specifications
Specifications
Gold Chloride Solution (0.1%)
125mL