Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
Supplier: Epredia™ SH87008
Description
- Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
- Background is stained green
Specifications
Silver Stain Kit, Periodic Acid |