Moldex™ Pesticide kit
Includes (1) 8000 series facepiece assembly, (5) 8910 N95 pre-filters, (1) 8020 filter retainer and (1 pair) SparkPlugs
Supplier:  Moldex™ 8113N
 View more versions of this product
Catalog No. 19-003343C
Explore available promotions
Description
Description
Specifications
Specifications
Paint Spray and Pesticide Applications which are Oil-free
Moldex™ 8000 Series Respirator
One 8000 Facepiece, one pair of 8100 Organic Vapors Cartridges, 8910 N95 Prefilters, one pair of 8020 Retainers, one pair of 6604 SparkPlugs™ foam Earplugs, Paint Bucket
Large