missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Acridine Orange
Cell-permeant nucleic acid binding dye
Supplier: Invitrogen™ A1301
Description
Acridine Orange emits green fluorescence when bound to dsDNA and red fluorescence when bound to ssDNA or RNA. This unique characteristic makes acridine orange useful for cell-cycle studies. Acridine orange has also been used as a lysosomal dye. This dye is available in solution (A-3568).
Apoptosis, Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, DNA Fragmentation, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing
Specifications
Orange | |
500⁄526 (DNA), 460⁄650 (RNA) | |
Fluorescence Microscope | |
Fluorescence | |
Fluorescent Dye | |
Nucleic Acids, Nucleus |
Store at room temperature and protect from light. | |
Cell-Permeant | |
1 g | |
Room Temperature | |
Dye |
For Research Use Only. Not for use in diagnostic procedures.