missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ BAPTA, Tetracesium Salt, cell impermeant
A cell-impermeant chelator, which is a highly selective for Ca2+ over Mg2+
Supplier: Invitrogen™ B1212
Description
- Can be used to control the level of extracellular Ca2+
- BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
Cell Viability & Proliferation | |
Chelator (Non-Fluorescent) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.